Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Dextran Sulfate, 50% Solution Sterile, OmniPur™, Calbiochem™,

Catalog No. 3730100ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-301-00ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-301-00ML Supplier MilliporeSigma™ Supplier No. 3730100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile-filtered, ready-to-use solution of dextran sulfate for molecular biology applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-18-1

Specifications

Color Tan to Yellow
Quantity 100 mL
Packaging Plastic Bottle
Grade Ungraded
Chemical Name or Material Dextran Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.