Learn More
Medchemexpress LLC Dextran sulfate low 100mg | 9011-18-1 | 40,000 g/mol | (C6H9NaO8S)n | 100 MG
SDP
Catalog No. 5000226562
Encompass_Preferred
Dextran sulfate, low sulfate (average MW ~40,000) is a sulfated polysaccharide supplied as a dry powder for research use. It is a polyanionic dextran derivative commonly used in biochemical assays, antiviral and anticoagulant studies, and as a cell-culture additive to reduce cell aggregation and support investigations into inflammation, fibrosis, and wound healing.
  • Low sulfate level for reduced charge density.
  • Average molecular weight of approximately 40,000 Da.
  • Polydisperse sulfated polysaccharide composed of repeating glucose units.
  • Supplied as a dry powder suitable for solution preparation.
  • Used as an anticoagulant and antiviral reagent in research assays.
  • Helps prevent cell aggregation when added to culture media.
Catalog No. 50-002-26562 Supplier Medchemexpress LLC Supplier No. HY116282M100MG
Dextran sulfate, low sulfate (average MW ~40,000) is a sulfated polysaccharide supplied as a dry powder for research use. It is a polyanionic dextran derivative commonly used in biochemical assays, antiviral and anticoagulant studies, and as a cell-culture additive to reduce cell aggregation and support investigations into inflammation, fibrosis, and wound healing.
  • Low sulfate level for reduced charge density.
  • Average molecular weight of approximately 40,000 Da.
  • Polydisperse sulfated polysaccharide composed of repeating glucose units.
  • Supplied as a dry powder suitable for solution preparation.
  • Used as an anticoagulant and antiviral reagent in research assays.
  • Helps prevent cell aggregation when added to culture media.