Learn More
Medchemexpress LLC Dextran sulfate low 10g | 9011-18-1 | 10 G
SDP
Catalog No. 5000227975
Encompass_Preferred
Dextran sulfate (sodium salt) is a sulfated polysaccharide research reagent (CAS 9011-18-1) with an average molecular weight of approximately 40,000 Da, supplied as a white to off-white powder in 10 g packaging. Consult the SDS and certificate of analysis for handling, storage, and composition details.
  • Sulfated polysaccharide reagent for research applications.
  • Average molecular weight ~40,000 Da.
  • White to off-white powder; store in a tight, dark container.
  • CAS number 9011-18-1.
  • Consult SDS and COA for composition and handling.
Catalog No. 50-002-27975 Supplier Medchemexpress LLC Supplier No. HY116282M10G
Dextran sulfate (sodium salt) is a sulfated polysaccharide research reagent (CAS 9011-18-1) with an average molecular weight of approximately 40,000 Da, supplied as a white to off-white powder in 10 g packaging. Consult the SDS and certificate of analysis for handling, storage, and composition details.
  • Sulfated polysaccharide reagent for research applications.
  • Average molecular weight ~40,000 Da.
  • White to off-white powder; store in a tight, dark container.
  • CAS number 9011-18-1.
  • Consult SDS and COA for composition and handling.