Learn More
Medchemexpress LLC Dextran sulfate low 1g | 9011-18-1 | 40,000 Da | (C6H9NaO8S)n | 1 G
SDP
Catalog No. 5000308045
Encompass_Preferred
Dextran sulfate, low sulfate (average MW 40,000) is a sulfated polysaccharide provided as the sodium salt for biochemical and cell-assay use. It is used experimentally as an anticoagulant and as a reagent with reported antiviral, antibacterial, anti-inflammatory, antifibrotic, and wound-healing activities.
  • Sulfated polysaccharide with average molecular weight ~40,000 Da.
  • Provided as the sodium salt, water-soluble powder.
  • Commonly used in biochemical and cell-based assays.
  • Reported activities: anticoagulant, antiviral, antibacterial, anti-inflammatory, antifibrotic, wound-healing.
  • Typically packaged in small research quantities; 1 G pack sizes are available.
Catalog No. 50-003-08045 Supplier Medchemexpress LLC Supplier No. HY116282M1G
Dextran sulfate, low sulfate (average MW 40,000) is a sulfated polysaccharide provided as the sodium salt for biochemical and cell-assay use. It is used experimentally as an anticoagulant and as a reagent with reported antiviral, antibacterial, anti-inflammatory, antifibrotic, and wound-healing activities.
  • Sulfated polysaccharide with average molecular weight ~40,000 Da.
  • Provided as the sodium salt, water-soluble powder.
  • Commonly used in biochemical and cell-based assays.
  • Reported activities: anticoagulant, antiviral, antibacterial, anti-inflammatory, antifibrotic, wound-healing.
  • Typically packaged in small research quantities; 1 G pack sizes are available.