Learn More
Medchemexpress LLC Dextran sulfate low 250mg | 9011-18-1 | 40,000 (average) | 250 MG
SDP
Catalog No. 5000226557
Encompass_Preferred
Dextran sulfate, low sulfate (MW 40,000) is a sulfated polysaccharide (dextran sulfate sodium salt) supplied as a laboratory reagent and cell culture additive to prevent cell aggregation. It is provided as an off-white to light-yellow powder and is reported to exhibit antiviral, antibacterial, anti-inflammatory, antifibrotic, and wound-healing activities.
  • Sulfated polysaccharide with average MW 40,000.
  • Low sulfate level compared with standard dextran sulfate grades.
  • Powder form, typically off-white to light-yellow.
  • Used as a cell culture additive to prevent cell aggregation.
  • Applicable in biochemical assays and as a gel-forming agent in formulations.
  • Available in laboratory pack sizes including 250 mg.
Catalog No. 50-002-26557 Supplier Medchemexpress LLC Supplier No. HY116282M250MG
Dextran sulfate, low sulfate (MW 40,000) is a sulfated polysaccharide (dextran sulfate sodium salt) supplied as a laboratory reagent and cell culture additive to prevent cell aggregation. It is provided as an off-white to light-yellow powder and is reported to exhibit antiviral, antibacterial, anti-inflammatory, antifibrotic, and wound-healing activities.
  • Sulfated polysaccharide with average MW 40,000.
  • Low sulfate level compared with standard dextran sulfate grades.
  • Powder form, typically off-white to light-yellow.
  • Used as a cell culture additive to prevent cell aggregation.
  • Applicable in biochemical assays and as a gel-forming agent in formulations.
  • Available in laboratory pack sizes including 250 mg.