Learn More
Medchemexpress LLC Dextran sulfate low 500mg | 9011-18-1 | 40,000 Da (average) | 500 MG
SDP
Catalog No. 5000308282
Encompass_Preferred
Dextran sulfate, low sulfate (average molecular weight ~40,000) is a sulfated polysaccharide used as a biochemical reagent. Common applications include reducing cell aggregation in culture, serving as a gel-forming agent in cosmetic formulations, and use in research probing complement/coagulation pathways and inflammatory or antiviral models.
  • Average molecular weight around 40,000 (polydisperse polymer).
  • Low-sulfate polyanion; soluble in aqueous media.
  • Used to reduce cell aggregation in cell culture applications.
  • Employs as a gel-forming or thickening agent in formulations.
  • Analytical profile typically lists water content, total sulfur, pH, and low heavy metals.
Catalog No. 50-003-08282 Supplier Medchemexpress LLC Supplier No. HY116282M500MG
Dextran sulfate, low sulfate (average molecular weight ~40,000) is a sulfated polysaccharide used as a biochemical reagent. Common applications include reducing cell aggregation in culture, serving as a gel-forming agent in cosmetic formulations, and use in research probing complement/coagulation pathways and inflammatory or antiviral models.
  • Average molecular weight around 40,000 (polydisperse polymer).
  • Low-sulfate polyanion; soluble in aqueous media.
  • Used to reduce cell aggregation in cell culture applications.
  • Employs as a gel-forming or thickening agent in formulations.
  • Analytical profile typically lists water content, total sulfur, pH, and low heavy metals.