Learn More
Medchemexpress LLC Dextran sulfate low 5g | 9011-18-1 | ca 40,000 Da | 5 G
SDP
Catalog No. 5000308056
Encompass_Preferred
Dextran sulfate (sodium salt) is a sulfated polysaccharide with an average molecular weight of approximately 40,000 Da (CAS 9011-18-1). Supplied as a dry powder for research use, it is used as a cell culture additive, a reagent in inflammation models, and as a biochemical inhibitor in coagulation and antiviral assays.
  • Average molecular weight around 40,000 Da.
  • Powder form for dissolution in aqueous buffers.
  • Reduces cell aggregation when used as a culture additive.
  • Used to induce colitis in animal models for inflammation studies.
  • Functions as an inhibitor of complement and coagulation pathways in assays.
Catalog No. 50-003-08056 Supplier Medchemexpress LLC Supplier No. HY116282M5G
Dextran sulfate (sodium salt) is a sulfated polysaccharide with an average molecular weight of approximately 40,000 Da (CAS 9011-18-1). Supplied as a dry powder for research use, it is used as a cell culture additive, a reagent in inflammation models, and as a biochemical inhibitor in coagulation and antiviral assays.
  • Average molecular weight around 40,000 Da.
  • Powder form for dissolution in aqueous buffers.
  • Reduces cell aggregation when used as a culture additive.
  • Used to induce colitis in animal models for inflammation studies.
  • Functions as an inhibitor of complement and coagulation pathways in assays.