Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran Sulfate (6000 to 8000), MP Biomedicals™

Catalog No. ICN10151850 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10151850 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10151850 Supplier MP Biomedicals, Inc Supplier No. 0210151850
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as an anticoagulant, antiviral and antilipemic agent

Dextran sulfate is a polyanionic derivative of dextran produced by esterification of Dextran with chlorosulphonic acid. Dextran sulfate sodium (DSS) is the sodium salt which is a white to off-white powder freely soluble in water and salt solutions to form a stable, clear solution.

  • Sulfur content is approximately 17% which corresponds to an average of 1.9 sulfate groups per glucosyl residue of the dextran molecule
  • High purity and consistent quality enables its application in pharmaceuticals and biotechnology industry
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Chemical Name or Material Dextran sulfate
pH 5.0 to 7.5 (1% aqueous solution)
Physical Form Powder
CAS 9011-18-1
Quantity 50 g
Molecular Formula (C6H7O5)m(C6H7O5)n
MDL Number MFCD00081551
Synonym dextran sulfate sodium, DSS
Solubility Information Soluble in water
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
IUPAC Name Dextran sulfate sodium salt
Molecular Weight (g/mol) 40,000
Formula Weight ≈1,400,000
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.