Learn More
Apexbio Technology LLC Dextran sulfate sodium salt,100g CAS# 9011-18-1
SDP
Supplier:  Apexbio Technology LLC B8205100000
Encompass_Preferred
The dextran sulfate sodium salt form makes it soluble and stable in water. Dextran sulfate contains approximately 17% sulfur, which corresponds to approximately 2.3 sulfate groups per glucose residue. Dextran sulfate sodium salt (DSS) is a polyanionic derivative of dextran formed by esterification of dextran and chlorosulfonic acid.Inflammatory bowel disease (IBD) is a chronic, relapsing gastrointestinal infection that increases the risk of intestinal tumors. In 1985, it first reported that dextran sodium salt (DSS) could be used to model hamster ulcerative colitis. A large number of studies have shown that the DSS colitis model is similar to human ulcerative colitis. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-223-9974