Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dextran sulfate sodi 100mg | 9011-18-1 | 100 MG
SDP

Catalog No. 5000228183
Encompass_Preferred

Dextran sulfate sodium salt (DSS) is a sulfated polysaccharide supplied as a powdered research reagent with a molecular weight range of 16,000-24,000 Da. It is used as a polyanionic biochemical reagent and to induce experimental colitis in animal models. Handle and store according to the material safety data sheet and supplier guidance.

  • Sulfated polysaccharide, polymeric structure.
  • Molecular weight range 16,000-24,000 Da.
  • Supplied as a powder; typical package 100 mg.
  • Used to induce experimental colitis and as a polyanionic reagent in biochemical assays.
  • Storage: powder -20 °C (up to 3 years) or 4 °C (up to 2 years); in solvent -80 °C (up to 6 months).
  • CAS number 9011-18-1.

Catalog No. 50-002-28183 Supplier Medchemexpress LLC Supplier No. HY116282B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dextran sulfate sodium salt (DSS) is a sulfated polysaccharide supplied as a powdered research reagent with a molecular weight range of 16,000-24,000 Da. It is used as a polyanionic biochemical reagent and to induce experimental colitis in animal models. Handle and store according to the material safety data sheet and supplier guidance.

  • Sulfated polysaccharide, polymeric structure.
  • Molecular weight range 16,000-24,000 Da.
  • Supplied as a powder; typical package 100 mg.
  • Used to induce experimental colitis and as a polyanionic reagent in biochemical assays.
  • Storage: powder -20 °C (up to 3 years) or 4 °C (up to 2 years); in solvent -80 °C (up to 6 months).
  • CAS number 9011-18-1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.