Learn More
Medchemexpress LLC Dextran sulfate sodi 10g | 9011-18-1 | 35,000-45,000 Da | 10 G
SDP
Catalog No. 5000227335
Encompass_Preferred
Dextran sulfate sodium salt is a sulfated polysaccharide (DSS) used in biomedical research, commonly to induce experimental colitis in rodent models. It is supplied as a sodium salt powder with a molecular weight range of 35,000-45,000 Da and is provided with batch-specific certificates of analysis and safety data sheets.
  • Molecular weight range 35,000-45,000 Da.
  • Polyanionic, water-soluble sodium salt form.
  • Used to induce colitis in in vivo inflammatory bowel disease models.
  • Provided as a white to off-white powder with COA and SDS documentation.
  • Suitable for research applications requiring a defined DSS molecular weight range.
Catalog No. 50-002-27335 Supplier Medchemexpress LLC Supplier No. HY116282C10G
Dextran sulfate sodium salt is a sulfated polysaccharide (DSS) used in biomedical research, commonly to induce experimental colitis in rodent models. It is supplied as a sodium salt powder with a molecular weight range of 35,000-45,000 Da and is provided with batch-specific certificates of analysis and safety data sheets.
  • Molecular weight range 35,000-45,000 Da.
  • Polyanionic, water-soluble sodium salt form.
  • Used to induce colitis in in vivo inflammatory bowel disease models.
  • Provided as a white to off-white powder with COA and SDS documentation.
  • Suitable for research applications requiring a defined DSS molecular weight range.