Learn More
Medchemexpress LLC Dextran sulfate sodi 10g | 9011-18-1 | 5,000 (average) Da | 10 G
SDP
Catalog No. 5000229490
Encompass_Preferred
Dextran sulfate sodium salt (CAS 9011-18-1) is a sulfated polysaccharide (sodium salt) used as a polyanionic reagent in biochemical and in vivo research. The MW 5,000 variant is supplied as a white to off-white powder and is commonly used for induction of experimental colitis and for assays affecting complement and coagulation pathways.
  • White to off-white powder.
  • Average molecular weight 5,000 Da.
  • Polyanionic reagent for biochemical and in vivo research.
  • Used to induce experimental colitis in animal models.
  • Inhibits complement and coagulation pathways in assays.
  • Available in multiple pack sizes, including 10 G.
  • Store powder at 4°C (2 years) or -20°C (3 years).
Catalog No. 50-002-29490 Supplier Medchemexpress LLC Supplier No. HY11628210G
Dextran sulfate sodium salt (CAS 9011-18-1) is a sulfated polysaccharide (sodium salt) used as a polyanionic reagent in biochemical and in vivo research. The MW 5,000 variant is supplied as a white to off-white powder and is commonly used for induction of experimental colitis and for assays affecting complement and coagulation pathways.
  • White to off-white powder.
  • Average molecular weight 5,000 Da.
  • Polyanionic reagent for biochemical and in vivo research.
  • Used to induce experimental colitis in animal models.
  • Inhibits complement and coagulation pathways in assays.
  • Available in multiple pack sizes, including 10 G.
  • Store powder at 4°C (2 years) or -20°C (3 years).