Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dextran sulfate sodi 1g | 9011-18-1 | 16,000-24,000 Da | 1 G
SDP

Catalog No. 5000226554
Encompass_Preferred

Dextran sulfate sodium salt (MW 16,000-24,000 Da) is a sulfated polysaccharide supplied as a powder for research use. It is commonly used to induce experimental colitis in animal models and can act as an inhibitor of viral adsorption. The material is provided as a sodium salt of sulfated dextran and ships at ambient temperature.

  • Sulfated polysaccharide, polymer of anhydroglucose.
  • Molecular weight approximately 16,000-24,000 Da.
  • CAS 9011-18-1.
  • Used to induce colitis models and inhibit viral adsorption.
  • Powder form; research use only; ships at ambient.
  • Available in 1 G pack sizes.

Catalog No. 50-002-26554 Supplier Medchemexpress LLC Supplier No. HY116282B1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dextran sulfate sodium salt (MW 16,000-24,000 Da) is a sulfated polysaccharide supplied as a powder for research use. It is commonly used to induce experimental colitis in animal models and can act as an inhibitor of viral adsorption. The material is provided as a sodium salt of sulfated dextran and ships at ambient temperature.

  • Sulfated polysaccharide, polymer of anhydroglucose.
  • Molecular weight approximately 16,000-24,000 Da.
  • CAS 9011-18-1.
  • Used to induce colitis models and inhibit viral adsorption.
  • Powder form; research use only; ships at ambient.
  • Available in 1 G pack sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.