Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dextran sulfate sodi 1g | 9011-18-1 | 35,000-45,000 Da | 1 G
SDP

Catalog No. 5000227336
Encompass_Preferred

Dextran sulfate sodium salt (MW 35,000-45,000) is a sulfated polysaccharide used primarily to induce experimental colitis in animal models. It is supplied as a sodium salt powder and is commonly used to model acute and chronic colitis and colitis-associated pathology.

  • Molecular weight 35,000-45,000 Da.
  • CAS number 9011-18-1.
  • Used to induce DSS-mediated colitis models in rodents.
  • Supplied as a sodium salt powder in a 1 g pack.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-27336 Supplier Medchemexpress LLC Supplier No. HY116282C1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dextran sulfate sodium salt (MW 35,000-45,000) is a sulfated polysaccharide used primarily to induce experimental colitis in animal models. It is supplied as a sodium salt powder and is commonly used to model acute and chronic colitis and colitis-associated pathology.

  • Molecular weight 35,000-45,000 Da.
  • CAS number 9011-18-1.
  • Used to induce DSS-mediated colitis models in rodents.
  • Supplied as a sodium salt powder in a 1 g pack.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.