Learn More
Medchemexpress LLC Dextran sulfate sodi 1g | 9011-18-1 | 450,000-550,000 Da | (C6H9NaO8S)n | 1 G
SDP
Catalog No. 5000308044
Encompass_Preferred
Dextran sulfate sodium salt (MW 450,000-550,000) is a high-molecular-weight sulfated polysaccharide supplied as a white to off-white solid for research use. It is commonly used in models of intestinal inflammation, antiviral studies, and in formulation of functional materials and nanoparticles.
  • Molecular weight range: 450,000-550,000 Da.
  • Sodium salt form supplied as a white to off-white solid.
  • Used to induce experimental colitis and for antiviral research (HIV-1, influenza A).
  • Applicable in nanoparticle formulation and functional material preparation.
  • Available in multiple package sizes, including 1 G.
  • Polymeric sulfated polysaccharide with formula (C6H9NaO8S)n.
Catalog No. 50-003-08044 Supplier Medchemexpress LLC Supplier No. HY116282D1G
Dextran sulfate sodium salt (MW 450,000-550,000) is a high-molecular-weight sulfated polysaccharide supplied as a white to off-white solid for research use. It is commonly used in models of intestinal inflammation, antiviral studies, and in formulation of functional materials and nanoparticles.
  • Molecular weight range: 450,000-550,000 Da.
  • Sodium salt form supplied as a white to off-white solid.
  • Used to induce experimental colitis and for antiviral research (HIV-1, influenza A).
  • Applicable in nanoparticle formulation and functional material preparation.
  • Available in multiple package sizes, including 1 G.
  • Polymeric sulfated polysaccharide with formula (C6H9NaO8S)n.