Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dextran sulfate sodi 1g | 9011-18-1 | 4500-5500 | 1 G
SDP

Catalog No. 5000227334
Encompass_Preferred

Dextran sulfate sodium salt (MW 4500-5500) is a sulfated polysaccharide provided as the sodium salt in powder form for laboratory research. It is commonly used in studies of coagulation, complement pathways, antiviral assays, and inflammatory disease models, and is supplied in small package sizes suitable for bench-top experiments.

  • Molecular weight range 4500-5500.
  • CAS number 9011-18-1.
  • Supplied as a white to off-white powder for laboratory use.
  • Functions as an anticoagulant and antiviral reagent in vitro.
  • Used in immune and inflammation model studies such as IBMIR inhibition.

Catalog No. 50-002-27334 Supplier Medchemexpress LLC Supplier No. HY116282A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dextran sulfate sodium salt (MW 4500-5500) is a sulfated polysaccharide provided as the sodium salt in powder form for laboratory research. It is commonly used in studies of coagulation, complement pathways, antiviral assays, and inflammatory disease models, and is supplied in small package sizes suitable for bench-top experiments.

  • Molecular weight range 4500-5500.
  • CAS number 9011-18-1.
  • Supplied as a white to off-white powder for laboratory use.
  • Functions as an anticoagulant and antiviral reagent in vitro.
  • Used in immune and inflammation model studies such as IBMIR inhibition.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.