Learn More
Medchemexpress LLC Dextran sulfate sodi 500mg | 9011-18-1 | 16,000-24,000 Da | 500 MG
SDP
Catalog No. 5000228403
Encompass_Preferred
Dextran sulfate sodium salt (MW 16,000-24,000) is a sulfated polysaccharide reagent used in research to modulate innate immune signaling, reported to induce STING polymerization and TBK1 activation. It is supplied as a white to off-white solid and is formulated for laboratory use with defined solubility and storage conditions.
  • Polymer average molecular weight 16,000-24,000.
  • White to off-white solid form.
  • High water solubility: 100 mg/mL (requires ultrasonic).
  • Low DMSO solubility: <1 mg/mL.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Intended for research use only.
Catalog No. 50-002-28403 Supplier Medchemexpress LLC Supplier No. HY116282B500MG
Dextran sulfate sodium salt (MW 16,000-24,000) is a sulfated polysaccharide reagent used in research to modulate innate immune signaling, reported to induce STING polymerization and TBK1 activation. It is supplied as a white to off-white solid and is formulated for laboratory use with defined solubility and storage conditions.
  • Polymer average molecular weight 16,000-24,000.
  • White to off-white solid form.
  • High water solubility: 100 mg/mL (requires ultrasonic).
  • Low DMSO solubility: <1 mg/mL.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Intended for research use only.