Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dextran sulfate sodi 500mg | 9011-18-1 | 4500-5500 Da | (C6H7NaO14S3)n | 500 MG
SDP

Catalog No. 5000229939
Encompass_Preferred

Dextran sulfate sodium salt (MW 4500-5500) is an anionic, sulfated polysaccharide (CAS 9011-18-1) used in biological research as a complement inhibitor, antiviral agent, and to induce colitis in experimental models. The material is supplied as a white to off-white powder, is water soluble, and is typically characterized by a molecular weight range of 4500-5500 Da and a purity of ≥90%.

  • Molecular weight range: 4500-5500 Da.
  • Appearance: white to off-white powder, soluble in water.
  • Purity: ≥90% as reported on the product datasheet.
  • Sulfate content typically 17.0-20.0% and specific optical rotation +95.0 to +105.0°.
  • Analytical properties: water content and loss on drying limits reported; heavy metals and endotoxin controlled.
  • Storage: keep frozen for long term or refrigerated in solution as specified by the datasheet.

Catalog No. 50-002-29939 Supplier Medchemexpress LLC Supplier No. HY116282A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dextran sulfate sodium salt (MW 4500-5500) is an anionic, sulfated polysaccharide (CAS 9011-18-1) used in biological research as a complement inhibitor, antiviral agent, and to induce colitis in experimental models. The material is supplied as a white to off-white powder, is water soluble, and is typically characterized by a molecular weight range of 4500-5500 Da and a purity of ≥90%.

  • Molecular weight range: 4500-5500 Da.
  • Appearance: white to off-white powder, soluble in water.
  • Purity: ≥90% as reported on the product datasheet.
  • Sulfate content typically 17.0-20.0% and specific optical rotation +95.0 to +105.0°.
  • Analytical properties: water content and loss on drying limits reported; heavy metals and endotoxin controlled.
  • Storage: keep frozen for long term or refrigerated in solution as specified by the datasheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.