Learn More
Medchemexpress LLC Dextran sulfate sodi 500mg | 9011-18-1 | 500 MG
SDP
Catalog No. 5000226555
Encompass_Preferred
Dextran sulfate sodium salt (MW 450000-550000) is a high-molecular-weight sulfated polysaccharide supplied as a white to off-white solid. It is used in biomedical research as an anionic polymer reagent (for example, neutrophil isolation) and in laboratory protocols that require a high-molecular-weight dextran sulfate. CAS 9011-18-1.
  • High molecular weight (450,000-550,000 g/mol).
  • Supplied as a white to off-white solid.
  • Available in multiple pack sizes, including 500 mg.
  • Used for neutrophil isolation and other laboratory procedures.
  • Anionic, water-soluble polysaccharide suitable for aqueous applications.
Catalog No. 50-002-26555 Supplier Medchemexpress LLC Supplier No. HY116282D500MG
Dextran sulfate sodium salt (MW 450000-550000) is a high-molecular-weight sulfated polysaccharide supplied as a white to off-white solid. It is used in biomedical research as an anionic polymer reagent (for example, neutrophil isolation) and in laboratory protocols that require a high-molecular-weight dextran sulfate. CAS 9011-18-1.
  • High molecular weight (450,000-550,000 g/mol).
  • Supplied as a white to off-white solid.
  • Available in multiple pack sizes, including 500 mg.
  • Used for neutrophil isolation and other laboratory procedures.
  • Anionic, water-soluble polysaccharide suitable for aqueous applications.