Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dextran sulfate sodi 5g | 9011-18-1 | 16,000-24,000 Da | 5 G
SDP

Catalog No. 5000308261
Encompass_Preferred

Dextran sulfate sodium salt (MW 16,000-24,000 Da) is a sulfated polysaccharide supplied as a dry powder for use as a biochemical reagent in assays and inflammation research. It is water soluble with sonication and should be stored refrigerated or frozen depending on storage duration.

  • Powder form for easy handling and weighing.
  • Molecular weight range 16,000-24,000 Da for consistent biological activity.
  • Soluble in water with sonication for stock preparation.
  • Available in convenient small-scale packaging for lab use.
  • Stable under refrigerated or frozen storage per datasheet.

Catalog No. 50-003-08261 Supplier Medchemexpress LLC Supplier No. HY116282B5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dextran sulfate sodium salt (MW 16,000-24,000 Da) is a sulfated polysaccharide supplied as a dry powder for use as a biochemical reagent in assays and inflammation research. It is water soluble with sonication and should be stored refrigerated or frozen depending on storage duration.

  • Powder form for easy handling and weighing.
  • Molecular weight range 16,000-24,000 Da for consistent biological activity.
  • Soluble in water with sonication for stock preparation.
  • Available in convenient small-scale packaging for lab use.
  • Stable under refrigerated or frozen storage per datasheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.