Learn More
Medchemexpress LLC Dextran sulfate sodi 5g | 9011-18-1 | 35,000-45,000 Da | (C6H7Na3O14S3)n | 5 G
SDP
Catalog No. 5000308260
Encompass_Preferred
Dextran sulfate sodium salt (MW 35,000-45,000) is a highly sulfated polysaccharide provided as a powder for laboratory research applications, commonly used to induce experimental colitis in rodent models and to study complement, coagulation, and antiviral responses.
  • High molecular weight fraction (35,000-45,000 Da) suitable for in vivo colitis models.
  • Water-soluble sulfated polysaccharide with biological activity in complement and coagulation assays.
  • Available in multiple laboratory pack sizes, including 5 g, for flexible experimental use.
  • Supplied as a dry powder with recommended storage conditions to maintain stability.
  • Applicable for in vitro and in vivo studies investigating inflammatory and antiviral mechanisms.
Catalog No. 50-003-08260 Supplier Medchemexpress LLC Supplier No. HY116282C5G
Dextran sulfate sodium salt (MW 35,000-45,000) is a highly sulfated polysaccharide provided as a powder for laboratory research applications, commonly used to induce experimental colitis in rodent models and to study complement, coagulation, and antiviral responses.
  • High molecular weight fraction (35,000-45,000 Da) suitable for in vivo colitis models.
  • Water-soluble sulfated polysaccharide with biological activity in complement and coagulation assays.
  • Available in multiple laboratory pack sizes, including 5 g, for flexible experimental use.
  • Supplied as a dry powder with recommended storage conditions to maintain stability.
  • Applicable for in vitro and in vivo studies investigating inflammatory and antiviral mechanisms.