Learn More
Medchemexpress LLC Dextran sulfate sodi 5g | 9011-18-1 | 450,000-550,000 | H2O4S.xNa.x | 5 G
SDP
Catalog No. 5000229853
Encompass_Preferred
Dextran sulfate sodium salt (MW 450,000-550,000) is a high-molecular-weight, sulfated dextran supplied as a white to off-white powder for laboratory research applications. It is used in biomedical studies (for example, induction of colitis in animal models), antiviral and anticoagulant assays, and materials research where sulfated polysaccharides are required.
  • White to off-white solid powder.
  • Molecular weight range 450,000-550,000.
  • Useful for induction of colitis models and antiviral research.
  • Analytical profile: water 6.2%, total sulfur 17.8%, free alkali ≤0.2%.
  • Available in small research pack sizes such as 5 G.
Catalog No. 50-002-29853 Supplier Medchemexpress LLC Supplier No. HY116282D5G
Dextran sulfate sodium salt (MW 450,000-550,000) is a high-molecular-weight, sulfated dextran supplied as a white to off-white powder for laboratory research applications. It is used in biomedical studies (for example, induction of colitis in animal models), antiviral and anticoagulant assays, and materials research where sulfated polysaccharides are required.
  • White to off-white solid powder.
  • Molecular weight range 450,000-550,000.
  • Useful for induction of colitis models and antiviral research.
  • Analytical profile: water 6.2%, total sulfur 17.8%, free alkali ≤0.2%.
  • Available in small research pack sizes such as 5 G.