Learn More
Medchemexpress LLC Dextran sulfate sodi 5g | 9011-18-1 | 4500-5500 | (C6H7Na3O14S3)n | 5 G
SDP
Catalog No. 5000227152
Encompass_Preferred
Dextran sulfate sodium salt (MW 4500-5500) is a sulfated polysaccharide supplied as the sodium salt powder for research use. It is used as an inhibitor of complement and coagulation pathways, as an anticoagulant and antiviral agent, and to induce experimental colitis in animal models. Store dry under recommended temperatures to maintain stability.
  • Molecular weight 4500-5500.
  • Available in laboratory-scale sizes, including 5 g.
  • Used to inhibit complement and coagulation pathways and as an anticoagulant.
  • Used in antiviral research and to induce experimental colitis models.
  • Powder form; store at -20°C (powder) or 4°C according to supplier recommendations.
Catalog No. 50-002-27152 Supplier Medchemexpress LLC Supplier No. HY116282A5G
Dextran sulfate sodium salt (MW 4500-5500) is a sulfated polysaccharide supplied as the sodium salt powder for research use. It is used as an inhibitor of complement and coagulation pathways, as an anticoagulant and antiviral agent, and to induce experimental colitis in animal models. Store dry under recommended temperatures to maintain stability.
  • Molecular weight 4500-5500.
  • Available in laboratory-scale sizes, including 5 g.
  • Used to inhibit complement and coagulation pathways and as an anticoagulant.
  • Used in antiviral research and to induce experimental colitis models.
  • Powder form; store at -20°C (powder) or 4°C according to supplier recommendations.