Learn More
Medchemexpress LLC Dextran sulfate sodi 5g | 9011-18-1 | 5 G
SDP
Catalog No. 5000227333
Encompass_Preferred
Dextran sulfate sodium salt (average MW ~5000) is a sulfated polysaccharide used as a research reagent. It functions as an inhibitor of complement and coagulation pathways and is employed in antiviral, anticoagulant, and immunology studies, including colitis models and cell-culture applications.
  • Average molecular weight approximately 5000
  • Polymeric, anionic sulfated dextran (repeating glucose units)
  • Assay reported as 99.2%
  • Solid, off-white to light yellow appearance
  • Available in small pack sizes suitable for laboratory use
Catalog No. 50-002-27333 Supplier Medchemexpress LLC Supplier No. HY1162825G
Dextran sulfate sodium salt (average MW ~5000) is a sulfated polysaccharide used as a research reagent. It functions as an inhibitor of complement and coagulation pathways and is employed in antiviral, anticoagulant, and immunology studies, including colitis models and cell-culture applications.
  • Average molecular weight approximately 5000
  • Polymeric, anionic sulfated dextran (repeating glucose units)
  • Assay reported as 99.2%
  • Solid, off-white to light yellow appearance
  • Available in small pack sizes suitable for laboratory use