Learn More
Medchemexpress LLC HY-P7223 10ug Medchemexpress, IL-6R (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501970586
Encompass_Preferred
Medchemexpress, HY-116282D 100mg Dextran sulfate sodium salt (MW 450000-550000) CAS:9011-18-1 Dextran sulfate sodium salt (MW 450000-550000) is a is a polymer of anhydroglucose with the molecular weight range of 450000-550000. Dextran sulfate sodium salt inhibits the replication of the human immunodeficiency virus by preventing the adsorption of the virus into host cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0586 Supplier Medchemexpress LLC Supplier No. HYP722310UG
Medchemexpress, HY-116282D 100mg Dextran sulfate sodium salt (MW 450000-550000) CAS:9011-18-1 Dextran sulfate sodium salt (MW 450000-550000) is a is a polymer of anhydroglucose with the molecular weight range of 450000-550000. Dextran sulfate sodium salt inhibits the replication of the human immunodeficiency virus by preventing the adsorption of the virus into host cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.