Learn More
Medchemexpress LLC HY-P7149 10ug Medchemexpress, CT-1 (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501970575
Encompass_Preferred
Medchemexpress, HY-116282A 100mg Dextran sulfate sodium salt (MW 4500-5500) CAS:9011-18-1 Dextran sulfate sodium salt (MW 4500-5500) is a is a polymer of anhydroglucose with the molecular weight range of 4500-5500. Dextran sulfate sodium salt inhibits the replication of the human immunodeficiency virus by preventing the adsorption of the virus into host cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0575 Supplier Medchemexpress LLC Supplier No. HYP714910UG
Medchemexpress, HY-116282A 100mg Dextran sulfate sodium salt (MW 4500-5500) CAS:9011-18-1 Dextran sulfate sodium salt (MW 4500-5500) is a is a polymer of anhydroglucose with the molecular weight range of 4500-5500. Dextran sulfate sodium salt inhibits the replication of the human immunodeficiency virus by preventing the adsorption of the virus into host cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.