Learn More
TARGETMOL CHEMICALS INC DEXTRAN SULFATE SODIUM 5000MG
SDP
Catalog No. 502577392
Encompass_Preferred
Also available in 500 mg 10000 mg and bulk. Please contact Fisher for quotes. Dextran sulfate sodium salt (MW 4500-5500) is an anhydrous glucose polymer with a molecular weight range of 4500-5500. Dextran sulfate sodium salt (MW 4500-5500) inhibits human immunodeficiency virus replication by preventing viral adsorption to host cells. Dextran sulfate sodium salt (MW 4500-5500) is a commonly used modeling compound for inflammatory bowel disease. It can destroy the integrity of the intestinal mucosal barrier through the high negative charge caused by sulfate groups leading to intestinal barrier damage macrophage dysfunction and bacterial flora disorder. purity: 98%
Catalog No. 50-257-7392 Supplier TARGETMOL CHEMICALS INC Supplier No. T136471G
Also available in 500 mg 10000 mg and bulk. Please contact Fisher for quotes. Dextran sulfate sodium salt (MW 4500-5500) is an anhydrous glucose polymer with a molecular weight range of 4500-5500. Dextran sulfate sodium salt (MW 4500-5500) inhibits human immunodeficiency virus replication by preventing viral adsorption to host cells. Dextran sulfate sodium salt (MW 4500-5500) is a commonly used modeling compound for inflammatory bowel disease. It can destroy the integrity of the intestinal mucosal barrier through the high negative charge caused by sulfate groups leading to intestinal barrier damage macrophage dysfunction and bacterial flora disorder. purity: 98%