Learn More
Medchemexpress LLC HY-P7219 10ug Medchemexpress, IL-4 (CHO-expressed), Porcine CAS: Purity:>98%
SDP
Catalog No. 501970585
Encompass_Preferred
Medchemexpress, HY-116282C 100mg Dextran sulfate sodium salt (MW 35000-45000) CAS:9011-18-1 Dextran sulfate sodium salt (MW 35000-45000) is a is a polymer of anhydroglucose with the molecular weight range of 35000-45000. Dextran sulfate sodium salt inhibits the replication of the human immunodeficiency virus by preventing the adsorption of the virus into host cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0585 Supplier Medchemexpress LLC Supplier No. HYP721910UG
Medchemexpress, HY-116282C 100mg Dextran sulfate sodium salt (MW 35000-45000) CAS:9011-18-1 Dextran sulfate sodium salt (MW 35000-45000) is a is a polymer of anhydroglucose with the molecular weight range of 35000-45000. Dextran sulfate sodium salt inhibits the replication of the human immunodeficiency virus by preventing the adsorption of the virus into host cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.