Learn More
Sigma Aldrich Fine Chemicals Biosciences Dextran sulfate sodium sal100G
Catalog No. 502902734
Encompass
Dextran sulfate is a negatively charged sulfated polysaccharide which is produced by the esterification of dextran with chlorosulphonic acid. It has antiviral anticoagulant and adjuvant properties. Dextran is used to treat hemorrhage burns surgery and trauma. It enhances blood flow and blocks erythrocyte aggregation. Dextran decreases the risk of thrombosis and postoperative pulmonary emboli. It acts as a ribonuclease inhibitor.
Catalog No. 50-290-2734 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D6001100G
Dextran sulfate is a negatively charged sulfated polysaccharide which is produced by the esterification of dextran with chlorosulphonic acid. It has antiviral anticoagulant and adjuvant properties. Dextran is used to treat hemorrhage burns surgery and trauma. It enhances blood flow and blocks erythrocyte aggregation. Dextran decreases the risk of thrombosis and postoperative pulmonary emboli. It acts as a ribonuclease inhibitor.