Learn More
Sigma Aldrich Fine Chemicals Biosciences Dextran sulfate sodium sal500G
Catalog No. 502914537
Encompass
Dextran sulfate is a negatively charged sulfated polysaccharide which is produced by the esterification of dextran with chlorosulphonic acid. It has antiviral anticoagulant and adjuvant properties. Dextran is used to treat hemorrhage burns surgery and trauma. It enhances blood flow and blocks erythrocyte aggregation. Dextran decreases the risk of thrombosis and postoperative pulmonary emboli. It acts as a ribonuclease inhibitor.
Catalog No. 50-291-4537 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D6001500G
Dextran sulfate is a negatively charged sulfated polysaccharide which is produced by the esterification of dextran with chlorosulphonic acid. It has antiviral anticoagulant and adjuvant properties. Dextran is used to treat hemorrhage burns surgery and trauma. It enhances blood flow and blocks erythrocyte aggregation. Dextran decreases the risk of thrombosis and postoperative pulmonary emboli. It acts as a ribonuclease inhibitor.