Learn More
Alkali Scientific Dextran Sulfate Sodium Salt, 250 G
SDP
Catalog No. 5000661845
Encompass_Preferred
Dextran Sulfate Sodium Salt is a sulfated polysaccharide commonly used in molecular biology and biochemistry research. This highly soluble reagent is primarily employed in cell culture applications, where it functions as an anticoagulant and a stabilizer in certain biochemical assays. It is particularly useful in studies involving blood coagulation, protein interactions, and the inhibition of virus binding. Dextran Sulfate is also used in a variety of laboratory applications such as gel filtration chromatography, cell culture medium supplementation, and as a reagent in the preparation of liposomes and other complex biological molecules. This 250g package is designed for use in molecular research and biochemistry laboratories that require high solubility and purity in their reagents.
Catalog No. 50-006-61845 Supplier Alkali Scientific Supplier No. DEX005CP
Dextran Sulfate Sodium Salt is a sulfated polysaccharide commonly used in molecular biology and biochemistry research. This highly soluble reagent is primarily employed in cell culture applications, where it functions as an anticoagulant and a stabilizer in certain biochemical assays. It is particularly useful in studies involving blood coagulation, protein interactions, and the inhibition of virus binding. Dextran Sulfate is also used in a variety of laboratory applications such as gel filtration chromatography, cell culture medium supplementation, and as a reagent in the preparation of liposomes and other complex biological molecules. This 250g package is designed for use in molecular research and biochemistry laboratories that require high solubility and purity in their reagents.