Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran Sulfate Sodium Salt (6,000-8,000 M.Wt), MP Biomedicals

Catalog No. MP210151890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210151890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210151890 Supplier MP Biomedicals, Inc Supplier No. 0210151890
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

  • Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research
  • DSS can be used as an anticoagulant, antiviral as well as antilipemic agent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name Dextran sulfate sodium salt
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White
Physical Form Powder
Solubility Information Soluble in water (100mg/mL - clear to slightly hazy yellow solution.
Quantity 500 g
Formula Weight 6,000-8,000
Chemical Name or Material DEXTRAN SULFATE SODIUM SALT (6,000-8,000 M.Wt)
Recommended Storage Store at Room Temperature (15-30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.