Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran Sulfate Sodium Salt, Colitis Grade (36,000 to 50,000), MP Biomedicals™

Catalog No. icn16011080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
50 g
100 g
500 g
1000 g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16011080 100 g
MP216011001 1 g
ICN16011010 10 g
MP216011025 25 g
ICN16011050 50 g
ICN16011090 500 g
02-160-11091 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. ICN16011080 Supplier MP Biomedicals, Inc Supplier No. 0216011080
Only null left
Add to Cart
Add to Cart

Induces severe colitis (inflammatory bowel disease) in laboratory animals

Dextran sulfate sodium salt, or DSS, is a synthetic sulfated branched polysaccharide derivative of dextran that has multiple uses in biomedical and clinical research. One important property of DSS is to trigger colitis in mice and rats when administered in drinking water. DSS binds to medium-chain-length fatty acids in the colon and induces intestinal inflammation. Colitis-grade DSS (MW = 36,000 to 50,000) from MP Bio is one of the most widely used products based on peer reviewed scientific publications.

  • Induces severe colitis (inflammatory bowel disease)
  • Highest sulfur content – 19%
  • Highest chirality - +104° of specific rotation
  • Lowest pH – 6.2 at 1% solution
TRUSTED_SUSTAINABILITY

Specifications

Color White
Chemical Name or Material Dextran Sulfate Sodium Salt
pH 5 to 7.11 (1% aqueous solution)
Physical Form Powder
CAS 9011-18-1
Boiling Point 310°C
Melting Point 92°C
Molecular Formula (C6H7O5)m(C6H7O5)n
MDL Number MFCD00081551
Solubility Information Soluble in water
Quantity 100 g
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
IUPAC Name Dextran sulfate sodium salt
Molecular Weight (g/mol) 40,000
For Use With (Application) Used for inducing ulcerative colitis in animals and extensively to study colon cancer developing in relation to colonic inflammation
Grade Colitis Grade
Moisture <10%
Packaging Plastic Container
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.