Learn More
Medchemexpress LLC Dextran sulfate sodium salt (MW 4500-5500) | 9011-18-1 | 4500-5500 | 10 G
SDP
Catalog No. 5000437816
Encompass_Preferred
Dextran sulfate sodium salt (DSS) is a polymer of dehydrated glucose with a molecular weight of approximately 4500-5500. It inhibits complement and coagulation pathways, belonging to the glycosaminoglycans (GAG) family. This compound functions as an anticoagulant, antiviral, and anti-lipemic agent, and can prevent HIV-1 virus adsorption to host cells, NK cell-mediated cytotoxicity, and instant blood-mediated inflammatory reaction (IBMIR).
  • Inhibitor of complement and coagulation pathways.
  • Belongs to the glycosaminoglycans (GAG) family.
  • Functions as an anticoagulant.
  • Acts as an antiviral agent.
  • Acts as an anti-lipemic agent.
  • Stops HIV-1 virus adsorption to host cells.
  • Prevents NK cell-mediated cytotoxicity.
  • Inhibits instant blood-mediated inflammatory reaction (IBMIR).
Catalog No. 50-004-37816 Supplier Medchemexpress LLC Supplier No. HY116282A10G
Dextran sulfate sodium salt (DSS) is a polymer of dehydrated glucose with a molecular weight of approximately 4500-5500. It inhibits complement and coagulation pathways, belonging to the glycosaminoglycans (GAG) family. This compound functions as an anticoagulant, antiviral, and anti-lipemic agent, and can prevent HIV-1 virus adsorption to host cells, NK cell-mediated cytotoxicity, and instant blood-mediated inflammatory reaction (IBMIR).
  • Inhibitor of complement and coagulation pathways.
  • Belongs to the glycosaminoglycans (GAG) family.
  • Functions as an anticoagulant.
  • Acts as an antiviral agent.
  • Acts as an anti-lipemic agent.
  • Stops HIV-1 virus adsorption to host cells.
  • Prevents NK cell-mediated cytotoxicity.
  • Inhibits instant blood-mediated inflammatory reaction (IBMIR).