Learn More
Dextran sulfate sodium salt, MW ca >500,000
Description
Dextran sulfate sodium is used to study colitis (inflammatory bowel disease) as well as an antiviral agent. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Yellow |
| Physical Form | Powder |
| Quantity | 50 g |
| Odor | Phenol-like |
| Merck Index | 14,2951 |
| Solubility Information | Soluble in water. |
| IUPAC Name | ethyl 4-aminobenzoate |
| Formula Weight | >≈500 kd |
| Chemical Name or Material | Dextran sulfate sodium salt |
Safety and Handling
| RTECSNumber | HH9290000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.