Learn More
Medchemexpress LLC Dextran T4 (MW 4,000) | 9004-54-0 | 3200-4800 | 10 G
SDP
Catalog No. 5000437548
Encompass_Preferred
Dextran 4,000 is a mucus rheology modifier that reduces mucus viscoelasticity and improves mucociliary/cough clearance. It is applicable for inhalation therapy or aerosol delivery in mucostatic respiratory diseases. Dextran is also a natural polysaccharide drug carrier that can be covalently bonded to drugs or self-assembled into nanoparticles and hydrogels. It is biodegradable, biocompatible, and enables targeted drug delivery and controlled release, with derivatives extending drug half-life and reducing immune clearance activity.
  • Reduces mucus cross-link density
  • Decreases mucus viscoelasticity
  • Improves mucociliary/cough clearance
  • Disrupts DNA-mucin network structure in mucus
  • Applicable for inhalation therapy in mucostatic respiratory diseases
  • Natural polysaccharide drug carrier
  • Biodegradable and biocompatible
  • Enables targeted drug delivery and controlled release
Catalog No. 50-004-37548 Supplier Medchemexpress LLC Supplier No. HY112624J10G
Add to Cart
Add to Cart
Dextran 4,000 is a mucus rheology modifier that reduces mucus viscoelasticity and improves mucociliary/cough clearance. It is applicable for inhalation therapy or aerosol delivery in mucostatic respiratory diseases. Dextran is also a natural polysaccharide drug carrier that can be covalently bonded to drugs or self-assembled into nanoparticles and hydrogels. It is biodegradable, biocompatible, and enables targeted drug delivery and controlled release, with derivatives extending drug half-life and reducing immune clearance activity.
  • Reduces mucus cross-link density
  • Decreases mucus viscoelasticity
  • Improves mucociliary/cough clearance
  • Disrupts DNA-mucin network structure in mucus
  • Applicable for inhalation therapy in mucostatic respiratory diseases
  • Natural polysaccharide drug carrier
  • Biodegradable and biocompatible
  • Enables targeted drug delivery and controlled release