Learn More
Medchemexpress LLC Dextran T7 (MW 7,000) | 9004-54-0 | 5600-8400 | 10 G
SDP
Catalog No. 5000437415
Encompass_Preferred
Dextran T7 (Dextran 7; Dextran D7) (MW 7,000) is a polymer of anhydroglucose with an average molecular weight of 7000. It exhibits good biodegradability and biocompatibility, making it suitable for use in food, pharmaceutics, cosmetics, and research.
  • Used as an encapsulation material for superparamagnetic magnetite nanoparticles, which can be formulated into a liver contrast agent for magnetic resonance imaging.
  • Can be conjugated with cytosine deaminase (CDA) from Aspergillus niger to enhance the enzyme's structural stability, resistance to protease degradation, and anti-proliferative activity.
Catalog No. 50-004-37415 Supplier Medchemexpress LLC Supplier No. HY112624T10G
Dextran T7 (Dextran 7; Dextran D7) (MW 7,000) is a polymer of anhydroglucose with an average molecular weight of 7000. It exhibits good biodegradability and biocompatibility, making it suitable for use in food, pharmaceutics, cosmetics, and research.
  • Used as an encapsulation material for superparamagnetic magnetite nanoparticles, which can be formulated into a liver contrast agent for magnetic resonance imaging.
  • Can be conjugated with cytosine deaminase (CDA) from Aspergillus niger to enhance the enzyme's structural stability, resistance to protease degradation, and anti-proliferative activity.