Learn More
Medchemexpress LLC Dextrin 500g
SDP
Catalog No. 5000206478
Encompass_Preferred
Dextrins are a group of low molecular weight carbohydrates produced by the hydrolysis of starch Dextrin is commonly used as a thickener stabilizer or binder in a variety of foods including baked goods beverages and confectionary In addition it is used in the production of adhesives paper and textiles Its unique chemical properties make it an important ingredient in a variety of industrial processes especially in construction and packaging
Catalog No. 50-002-06478 Supplier Medchemexpress LLC Supplier No. HYW115731500G
Dextrins are a group of low molecular weight carbohydrates produced by the hydrolysis of starch Dextrin is commonly used as a thickener stabilizer or binder in a variety of foods including baked goods beverages and confectionary In addition it is used in the production of adhesives paper and textiles Its unique chemical properties make it an important ingredient in a variety of industrial processes especially in construction and packaging