Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrin, Commercial grade Type I, MP Biomedicals™

Catalog No. ICN10151790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN10151790 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10151790 Supplier MP Biomedicals, Inc Supplier No. 0210151790

Specifications

Color White to Yellow
Physical Form Powder
CAS 9004-53-9
Molecular Formula C18H32O16
MDL Number MFCD00081554
Density 1.45g/mL (L)
Synonym dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
Quantity 500 g
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
IUPAC Name (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 504.438
PubChem CID 62698
Grade Commercial
Chemical Name or Material Dextrin, For Nutrition Studies
Show More Show Less
Recommended Storage Store at Room Temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.