Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrin, White Powder, Spectrum™ Chemical
SDP

Catalog No. 18602100 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-100 50 kg Poly Bottle
18-602-099 2.5 kg Poly Bottle
18-602-098 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-100 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DE13750KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DE137, 9004-53-9, (C6H10O5)n·xH2O

Spectrum™ Chemical Dextrin, White Powder , is used as water-soluble glue in envelope adhesives, crispness enhancers in food processing and as a thickening agent in pharmaceuticals. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9004-53-9
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.44
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
IUPAC Name (3R,4S,5S,6R)-2-{[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](O)C(O[C@H]3[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 100%
Quantity 50 kg
Packaging Poly Bottle
Grade Ungraded
Chemical Name or Material Dextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.