Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrinized Corn Starch, MP Biomedicals

Catalog No. MP296042905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 lb.
25 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP296042905 5 lb.
MP296042925 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP296042905 Supplier MP Biomedicals, Inc Supplier No. 0296042905
Only null left
Add to Cart
Add to Cart

  • A component in animal diet

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym Maltodextin

Specifications

Color White
Quantity 5 lb.
Physical Form Powder
Chemical Name or Material DEXTRINIZED CORN STARCH
Recommended Storage Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.