Learn More
Waters Corp Dextro-Pak Cartridge Column, 8.0 x 100 mm, Reversed-Phase, Carbohydrate Analysis
Catalog No. 502391959 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Dextro-Pak Cartridge Column is designed for the analysis of sugars, carbohydrates, organic acids, and alcohols. It operates in reversed phase mode and is suitable for various applications, including the analysis of hydrolysed syrups and derivatized sugars.
  • Designed for the analysis of sugars, carbohydrates, organic acids, and alcohols
  • Operates in reversed phase mode
  • Dimensions of 8.0 × 100 mm
  • Used for hydrolysed syrups and derivatized sugars
  • Smallest elute first during analysis
Catalog No. 50-239-1959 Supplier Waters Corp Supplier No. WAT085650
Add to Cart
Add to Cart
The Dextro-Pak Cartridge Column is designed for the analysis of sugars, carbohydrates, organic acids, and alcohols. It operates in reversed phase mode and is suitable for various applications, including the analysis of hydrolysed syrups and derivatized sugars.
  • Designed for the analysis of sugars, carbohydrates, organic acids, and alcohols
  • Operates in reversed phase mode
  • Dimensions of 8.0 × 100 mm
  • Used for hydrolysed syrups and derivatized sugars
  • Smallest elute first during analysis