Learn More
American Radiolabeled Chemicals Inc DEXTROMETH 14C 50UC
SDP
Catalog No. 50901254
Encompass_Preferred
Dextromethorphan-N-methyl-14C 50uCi, 50-60 mCi/mmol, ethanol, 0.1 mCi/ml, M.W. 271.41, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-254 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 343950UC
Dextromethorphan-N-methyl-14C 50uCi, 50-60 mCi/mmol, ethanol, 0.1 mCi/ml, M.W. 271.41, Shipped in Dry Ice, Exclusive