Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextromethorphan HBr Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015123 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5123 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5123 Supplier MilliporeSigma Supelco Supplier No. PHR1018500MG
Add to Cart
Add to Cart
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Dextromethorphan HBr is a cough suppressant and a mucolytic agent, widely used in the treatment of various respiratory disorders such as for the relief of non-productive cough. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 6700-34-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C18H25 NO · HBr · H2O
MDL Number MFCD02173901
Synonym Dextromethorphan hydrobromide monohydrate; (9 S,13 S,14 S)-3-Methoxy-17-methylmorphinan hydrobromide
Type Certified Reference Material
Quantity 500 mg
Formula Weight 370.32
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.