Learn More
Dextromethorphan N-Oxide Hydrochloride, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 1177419-85-0 |
| Flash Point | Not Applicable |
| Molecular Formula | C18H25NO2 · HCl |
| Linear Formula | C18H25NO2 · HCl |
| Quantity | 100 mg |
| Synonym | (1 S,9 S,10 S)-4-Methoxy-17-methyl-17-oxido-17-azoniatetracyclo[7.5.3.01,10.02,7 ]heptadeca-2(7),3,5-triene hydrochloride |
| Molecular Weight (g/mol) | 323.86 |
| Formula Weight | 323.86 |
| Grade | Certified Reference Material |
| Physical Form | Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.