Learn More
Cayman Chemical Dextromethorphan, Purity: ≥98%, CAS: 125-71-3, 50 mg
SDP
Catalog No. 502952398
Encompass_Preferred
An analytical reference standard categorized as an antitussive; has analgesic and anticonvulsant activities in rodents; has been found as an adulterant in 3,4-MDMA; has potential for abuse; intended for research and forensic applications
Catalog No. 50-295-2398 Supplier Cayman Chemical Supplier No. 1115750 MG
An analytical reference standard categorized as an antitussive; has analgesic and anticonvulsant activities in rodents; has been found as an adulterant in 3,4-MDMA; has potential for abuse; intended for research and forensic applications