Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bound Tree Medical Dextrose, 5328-65-4, MFCD00063684, 500 mL Bag

Catalog No. NC1493403
Encompass_Preferred

Molecular Formula: C6 H12 O6, Molecular Weight: 180.155, 5%.

Catalog No. NC1493403 Supplier Bound Tree Medical Supplier No. 600063CS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula: C6 H12 O6, Molecular Weight: 180.155, 5%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.