Learn More
Med Vet International 5% Dextrose Injection, 1000mL
Catalog No. 502827830
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dextrose 5% in water Inj USP LifeCare are sterile and nonpyrogenic. They are large volume parenteral solutions containing various concentrations and combinations of these drugs in water for injections intended for intravenous administration. This solution contains no bacteriostat, antimicrobial agent or buffer and each is intended only as a single-dose injection. When smaller doses are required the unused portion should be discarded. Freight will be charged at actual freight cost.
Catalog No. 50-282-7830 Supplier Med Vet International Supplier No. RXDEXWATEREA
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dextrose 5% in water Inj USP LifeCare are sterile and nonpyrogenic. They are large volume parenteral solutions containing various concentrations and combinations of these drugs in water for injections intended for intravenous administration. This solution contains no bacteriostat, antimicrobial agent or buffer and each is intended only as a single-dose injection. When smaller doses are required the unused portion should be discarded. Freight will be charged at actual freight cost.