Learn More
Valley Vet Supply DEXTROSE 50% 500ML
Catalog No. NC1941356
Encompass
Dextrose 50% for Animal Use 500 MLUsed in treatment of acetonemia or ketosis in cattle and as a source of carbohydrates. Dosage: 50 cc to 100 lbs body weight IV.
Catalog No. NC1941356 Supplier Valley Vet Supply Supplier No. 16035
Dextrose 50% for Animal Use 500 MLUsed in treatment of acetonemia or ketosis in cattle and as a source of carbohydrates. Dosage: 50 cc to 100 lbs body weight IV.