Learn More
U.S. Pharmacopeia Dextrose, 50-99-7, MFCD00148912, 500mg
Catalog No. NC9024327
Encompass
Molecular formula C6H12O6, Molecular weight 180.2, Melting Point 294.8 °F (146 °C), Synonyms: Corn sugar, Glucose
Catalog No. NC9024327 Supplier U.S. Pharmacopeia Supplier No. 1181302
Molecular formula C6H12O6, Molecular weight 180.2, Melting Point 294.8 °F (146 °C), Synonyms: Corn sugar, Glucose